Learn More
Teknova LB Agar Plates with Gent-7, Kan-50, Tet-10, X-Gal, IPTG. 100x100mm, 20 Plates/Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387713
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 7g/mL Gentamicin Sulfate, 50g/mL Kanamycin Sulfate,10g/mL Tetracycline Hydrochloride, 60g/mL X-Gal,0.1mM Isopropyl-B-D-thiogalactopyranoside(IPTG)
Catalog No. 50-238-7713 Supplier Teknova Supplier No. L39188PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 7g/mL Gentamicin Sulfate, 50g/mL Kanamycin Sulfate,10g/mL Tetracycline Hydrochloride, 60g/mL X-Gal,0.1mM Isopropyl-B-D-thiogalactopyranoside(IPTG)