Learn More
Teknova LB Agar Plates with Hygromycin-100, 100mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387653
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100g/mL Hygromycin
Catalog No. 50-238-7653 Supplier Teknova Supplier No. L10568PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100g/mL Hygromycin