Learn More
Teknova LB Agar Plates with IPTG and X-Gal. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387568
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,60g/mL X-Gal, 0.1mM Isopropyl-B-D-thiogalactopyranoside(IPTG)
Catalog No. 50-238-7568 Supplier Teknova Supplier No. L19204PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,60g/mL X-Gal, 0.1mM Isopropyl-B-D-thiogalactopyranoside(IPTG)