Learn More
Teknova LB Agar Plates with Kanamycin-100. 100mm, 10 Plates per Sleeve, Sterile. 4 Pack.
SDP
Catalog No. 502387633
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Kanamycin Sulfate
Catalog No. 50-238-7633 Supplier Teknova Supplier No. L51274PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Kanamycin Sulfate