Learn More
Teknova LB Agar Plates with Kan-35, 100mm, 20 Plates per Sleeve, Sterile 4 pack.
SDP
Catalog No. 502387389
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 35g/mL Kanamycin Sulfate
Catalog No. 50-238-7389 Supplier Teknova Supplier No. L10434PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 35g/mL Kanamycin Sulfate