Learn More
Teknova LB Agar Plates with Kanamycin-50, X-Gal, and IPTG. 100mm, 20 Plates per Sleeve, Sterile. 4 Pack.
SDP
Catalog No. 502387834
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 50g/mL Kanamycin Sulfate, 60g/mL X-gal, 0.1mM Isopropyl-B-D-thiogalactopyranoside(IPTG)
Catalog No. 50-238-7834 Supplier Teknova Supplier No. L19094PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 50g/mL Kanamycin Sulfate, 60g/mL X-gal, 0.1mM Isopropyl-B-D-thiogalactopyranoside(IPTG)