Learn More
Teknova LB Agar Plates with Kanamycin-20. 100mm, 20 Plates per Sleeve, Sterile 4 pack.
SDP
Supplier:  Teknova L10794PK
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 20g/mL Kanamycin Sulfate
Catalog No. 50-238-7678