Learn More
Teknova LB Agar Plates with Kanamycin-50. 200mL Fill, 245X245mm, 4 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387914
Encompass
1.0% Tryptone, 0.5% Yeast Extract,1.0% Sodium Chloride, 1.5% Agar, 50g/mL Kanamycin Sulfate
Catalog No. 50-238-7914 Supplier Teknova Supplier No. L60504PK
1.0% Tryptone, 0.5% Yeast Extract,1.0% Sodium Chloride, 1.5% Agar, 50g/mL Kanamycin Sulfate