Learn More
Teknova LB Agar Plates with Kanamycin-50. 86X128mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502388125
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Kanamycin Sulfate
Catalog No. 50-238-8125 Supplier Teknova Supplier No. L20254PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Kanamycin Sulfate