Learn More
Teknova LB Agar Plates with Kanamycin-50. 100mm, 10 Plates per Sleeve, Sterile.
SDP
Catalog No. 502387991
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/ml Kanamycin Sulfate
Catalog No. 50-238-7991 Supplier Teknova Supplier No. L1155
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/ml Kanamycin Sulfate
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.