Learn More
Teknova LB Agar Plates with Kanamycin-50. 245x245mm Corning Low Profile, 4 Plates per Sleeve, Sterile
SDP
Catalog No. 502387996
Encompass
1.0% Tryptone, 0.5% Yeast Extract,1.0% Sodium Chloride, 1.5% Agar,50g/mL Kanamycin Sulfate
Catalog No. 50-238-7996 Supplier Teknova Supplier No. L6025
1.0% Tryptone, 0.5% Yeast Extract,1.0% Sodium Chloride, 1.5% Agar,50g/mL Kanamycin Sulfate