Learn More
Teknova LB Agar Plates with Kanamycin-50. 150mm, 20 Plates per Sleeve, Sterile.
SDP
Supplier:  Teknova L5025
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Kanamycin Sulfate
Catalog No. 50-238-8196