Learn More
Teknova Agar Plates with Kanamycin-50. 100mm, 20 Plates per Sleeve, Sterile
SDP
Catalog No. 502388200
Encompass
LB Agar Plates with Kanamycin-25 are a nutrient medium that supports the growth and selection of kanamycin-resistant E. coli strains. The inclusion of 25 µg/mL kanamycin sulfate enables selection and maintenance of plasmid-bearing transformants on solid LB.
Requires refrigeration upon delivery
1.0% Tryptone
0.5% Yeast Extract
1.0% Sodium Chloride
1.5% Agar
25µg/mL Kanamycin Sulfate
Catalog No. 50-238-8200 Supplier Teknova Supplier No. L1025
LB Agar Plates with Kanamycin-25 are a nutrient medium that supports the growth and selection of kanamycin-resistant E. coli strains. The inclusion of 25 µg/mL kanamycin sulfate enables selection and maintenance of plasmid-bearing transformants on solid LB.
Requires refrigeration upon delivery
1.0% Tryptone
0.5% Yeast Extract
1.0% Sodium Chloride
1.5% Agar
25µg/mL Kanamycin Sulfate