Learn More
Teknova LB Agar Plates with Kanamycin-100. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388127
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100g/mL Kanamycin Sulfate
Catalog No. 50-238-8127 Supplier Teknova Supplier No. L1027
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100g/mL Kanamycin Sulfate