Learn More
Teknova LB Agar Plates with Kanamycin-50. 100mm Plates, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388194
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 50g/mL Kanamycin Sulfate
Catalog No. 50-238-8194 Supplier Teknova Supplier No. L7725
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 50g/mL Kanamycin Sulfate