Learn More
Teknova LB Agar Plates with Kanamycin-40. 100mm Plates, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388201
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,40g/mL Kanamycin Sulfate
Catalog No. 50-238-8201 Supplier Teknova Supplier No. L1053
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,40g/mL Kanamycin Sulfate