Learn More
Teknova LB Agar Plates with Kanamycin-50, Tetracycline-10. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387554
Encompass
1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Kanamycin Sulfate, 10g/mL Tetracycline Hydrochloride
Catalog No. 50-238-7554 Supplier Teknova Supplier No. L13684PK
1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Kanamycin Sulfate, 10g/mL Tetracycline Hydrochloride