Learn More
Teknova LB Agar Plates with Kanamycin-50 and Streptomycin-50, 100mm, 20 plates/sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387528
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 50g/mL Kanamycin Sulfate, 50g/mL Streptomycin
Catalog No. 50-238-7528 Supplier Teknova Supplier No. L13268PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 50g/mL Kanamycin Sulfate, 50g/mL Streptomycin