Learn More
Teknova LB Agar Plates with Kanamycin-30. 150mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387268
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,30g/mL Kanamycin Sulfate
Catalog No. 50-238-7268 Supplier Teknova Supplier No. L50244PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,30g/mL Kanamycin Sulfate