Learn More
Teknova LB Agar Plates with Kanamycin-200. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387318
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 200g/mL Kanamycin Sulfate
Catalog No. 50-238-7318 Supplier Teknova Supplier No. L10774PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 200g/mL Kanamycin Sulfate