Learn More
Teknova LB Agar Plates with Streptomycin-50. 100mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387408
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Streptomycin Sulfate
Catalog No. 50-238-7408 Supplier Teknova Supplier No. L10298PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Streptomycin Sulfate