Learn More
Teknova LB Agar Plates with Tetracycline-10. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387622
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,10g/mL Tetracycline Hydrochloride
Catalog No. 50-238-7622 Supplier Teknova Supplier No. L10324PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,10g/mL Tetracycline Hydrochloride