Learn More
Teknova LB Agar Plates with Tetracycline-5. 100mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387454
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 5g/mL Tetracycline Hydrochloride
Catalog No. 50-238-7454 Supplier Teknova Supplier No. L11338PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 5g/mL Tetracycline Hydrochloride