Learn More
Teknova LB Agar Plates with Tetracycline-25. 100mm, 20 plates/sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387911
Encompass
1.0% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 1.5% Agar,25g/mL Tetracycline Hydrochloride
Catalog No. 50-238-7911 Supplier Teknova Supplier No. L10354PK
1.0% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 1.5% Agar,25g/mL Tetracycline Hydrochloride