Learn More
Teknova LB Agar Plates with Tetracycline-12.5. 100mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387317
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 12.5g/mL Tetracycline Hydrochloride
Catalog No. 50-238-7317 Supplier Teknova Supplier No. L10678PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 12.5g/mL Tetracycline Hydrochloride