Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals LB Agar plate with 50μg/mL Spectinomycin

Catalog No. aaj60541eqf
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60541EQF 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60541EQF Supplier Thermo Scientific Chemicals Supplier No. J60541EQF
Only null left
Add to Cart

 

It is used in the resistance of 16S ribosomal RNA in Pasteurella multocida.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Used for growth of Escherichia coli, plasmid and other bacterial cultures
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Form Solid
Media Type LB Agar
For Use With (Application) For growth of Escherichia coli, plasmid and other bacterial cultures
Packaging Type Plate
Quantity 20 Plates
Height (Metric) 15 mm
Diameter (Metric) Outside 10 cm

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.