Learn More
Crescent Chemical Co Inc LB AGAR POWDER 35G FOR 1L
SDP
Catalog No. 50249150
Encompass_Preferred
LB Agar, Powder 35 g/L For cultivation of E. coli in molecular biology.10 g/l Tryptone5 g/l Yeast extract5 g/l NaCl15 g/l AgarFor making 1 L liquid medium suspend 35 g in 900 ml distilled water, adjust the pH to 7.0 with approximately 0.2 ml of 5N NaOH, fill up t
Catalog No. 50-249-150 Supplier Crescent Chemical Co Inc Supplier No. SE48502.01
LB Agar, Powder 35 g/L For cultivation of E. coli in molecular biology.10 g/l Tryptone5 g/l Yeast extract5 g/l NaCl15 g/l AgarFor making 1 L liquid medium suspend 35 g in 900 ml distilled water, adjust the pH to 7.0 with approximately 0.2 ml of 5N NaOH, fill up t