Learn More
Teknova LB Agar Plates with Carbenicillin-100 and Chloramphenicol-34. 100mm, 20 Plates/Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387925
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100g/mL Carbenicillin Disodium Salt, 34g/mL Chloramphenicol
Catalog No. 50-238-7925 Supplier Teknova Supplier No. L27608PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100g/mL Carbenicillin Disodium Salt, 34g/mL Chloramphenicol