Learn More
MOLTOX Molecular Toxicology Inc LB Agar, Lennox L w/ 40 ug/ml Kanamycin, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948843
Encompass_Preferred
LB Agar, Lennox L, also referred to as Luria-Bertani Agar, is a widely used medium in molecular biology applications for maintaining and cultivating recombinant strains of E. coli. LB Agar, Lennox contains half the amount of sodium chloride as that of the Miller formulation which allows for the researcher to be selective of the salt content when working with a specific strain. MOLTOX offers a wide variety of LB Agar, Lennox plates with the addition of several different types and concentrations of antibiotics to provide a more selective medium for recombinant cells with a specific antibiotic resistance. Used as an industry standard for maintaining and cultivating recombinant strains of E. coli and for the preparation of plasmid DNA, MOLTOX offers a large selection of LB Agars and Broths with plenty of added antibiotics to choose from for a more selective media. Some media are temperature sensitive and may have specific temperature and storage requirements for shipping/delivery.
Catalog No. 50-948-843 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S114
LB Agar, Lennox L, also referred to as Luria-Bertani Agar, is a widely used medium in molecular biology applications for maintaining and cultivating recombinant strains of E. coli. LB Agar, Lennox contains half the amount of sodium chloride as that of the Miller formulation which allows for the researcher to be selective of the salt content when working with a specific strain. MOLTOX offers a wide variety of LB Agar, Lennox plates with the addition of several different types and concentrations of antibiotics to provide a more selective medium for recombinant cells with a specific antibiotic resistance. Used as an industry standard for maintaining and cultivating recombinant strains of E. coli and for the preparation of plasmid DNA, MOLTOX offers a large selection of LB Agars and Broths with plenty of added antibiotics to choose from for a more selective media. Some media are temperature sensitive and may have specific temperature and storage requirements for shipping/delivery.