Learn More
Teknova LB Agarose Plates with Kanamycin-50. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387370
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agarose, 50g/mL Kanamycin Sulfate
Catalog No. 50-238-7370 Supplier Teknova Supplier No. L55154PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agarose, 50g/mL Kanamycin Sulfate