Learn More
Kd Medical Inc LB Ampicillin plates 100mm, 100ug/ml.
SDP
Catalog No. 501003477
Encompass_Preferred
LB Ampicillin plates. Used for the maintenance and propagation of Escheria coli (E.coli), selective for Ampicillin resistance. Ten 100mm plates per sleeve. Concentration of Ampicillin is 100ug/ml.
Catalog No. 50-100-3477 Supplier Kd Medical Inc Supplier No. BPL2200
LB Ampicillin plates. Used for the maintenance and propagation of Escheria coli (E.coli), selective for Ampicillin resistance. Ten 100mm plates per sleeve. Concentration of Ampicillin is 100ug/ml.