Learn More
Kd Medical Inc LB+ AMP 100UG/ML XGAL 40UG/ML
SDP
Supplier:  Kd Medical Inc BPL2310
Encompass_Preferred
Used for the maintenance and propagation of Escheria coli (E.coli), selective for Ampicillin (100ug/ml) resistance. X-Gal (40ug/ml) used for blue/white screening of colonies. IPTG .5mM, Ten 100mm plates per sleeve.
Catalog No. 50-101-8915