Learn More
Kd Medical Inc LB AMP 50MG/ML
SDP
Catalog No. 501003476
Encompass_Preferred
LB Ampicillin plates. Used for the maintenance and propagation of E. coli, selective for ampicillin resistance. Ten 100mm plates per sleeve. Concentration of ampicillin is 50 mg/ml.
Catalog No. 50-100-3476 Supplier Kd Medical Inc Supplier No. BPL2070
LB Ampicillin plates. Used for the maintenance and propagation of E. coli, selective for ampicillin resistance. Ten 100mm plates per sleeve. Concentration of ampicillin is 50 mg/ml.