Learn More
Kd Medical Inc LB + AMP + XGAL + IPTG
SDP
Catalog No. 501018986
Encompass_Preferred
LB Ampicillin 75ug/ml, Xgal 40ug/ml, IPTG .5mM plates are used for the maintenance and propagation of Escheria coli (E.coli), selective for Ampicillin resistance. X-gal is used for the blue/white screening of colonies.
Catalog No. 50-101-8986 Supplier Kd Medical Inc Supplier No. BPL2170
LB Ampicillin 75ug/ml, Xgal 40ug/ml, IPTG .5mM plates are used for the maintenance and propagation of Escheria coli (E.coli), selective for Ampicillin resistance. X-gal is used for the blue/white screening of colonies.