Learn More
Kd Medical Inc LB + AMP+ XGAL+ IPTG
SDP
Supplier:  Kd Medical Inc BPL2310C
Encompass
Used for the maintenance and propagation of Escheria coli (E.coli), selective for Ampicillin (100ug/ml) resistance. X-Gal (80ug/ml) used for blue/white screening of colonies. IPTG .5mM, Ten 100mm plates per sleeve.
Catalog No. 50-101-8998