Learn More
Teknova LB Animal-Free Agar. 500mL, Sterile.
SDP
Catalog No. 502388055
Encompass
1.69% Animal-Free Soy Peptone,0.50% Yeast Extract, 1.00% Sodium Chloride, 1.50% Agar
Catalog No. 50-238-8055 Supplier Teknova Supplier No. L8072
1.69% Animal-Free Soy Peptone,0.50% Yeast Extract, 1.00% Sodium Chloride, 1.50% Agar