Learn More
LAMDA BIOTECH INC LB Broth
Catalog No. 502045826
Encompass_Preferred
Premixed LB powder: weigh it out and put it in the flask or bottle. Add the appropriate volume of water and autoclave as usual.
Catalog No. 50-204-5826 Supplier LAMDA BIOTECH INC Supplier No. C120
Premixed LB powder: weigh it out and put it in the flask or bottle. Add the appropriate volume of water and autoclave as usual.