Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp LB Broth 20 Gram Buffered Capsules [Lennox L Broth], 1 Kilogram
SDP

Catalog No. 50550421
Encompass_Preferred

RPI media is now available in convenient premeasured and pre-mixed capsules. Prebuffered media eliminates pH adjustments, dust and smells from the lab. Low Salt Formula. Molecular Biology Grade.

For propagation and maintenance of E.Coli:

Add 1 capsule per liter of water.
Heat and stir until capsule is completely dissolved.
Buffer is set for 7.2 at 20°C using a 10mM solution of TRIS/TRIS HCl
Sterilize by autoclaving for 15 minutes.

Ingredients:
Casein Digest of Peptone: 10 g/L
Yeast Extract: 5 g/L
Sodium Chloride: 5 g/L

Catalog No. 50-550-421 Supplier Research Products International Corp Supplier No. L240651000.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RPI media is now available in convenient premeasured and pre-mixed capsules. Prebuffered media eliminates pH adjustments, dust and smells from the lab. Low Salt Formula. Molecular Biology Grade.

For propagation and maintenance of E.Coli:

Add 1 capsule per liter of water.
Heat and stir until capsule is completely dissolved.
Buffer is set for 7.2 at 20°C using a 10mM solution of TRIS/TRIS HCl
Sterilize by autoclaving for 15 minutes.

Ingredients:
Casein Digest of Peptone: 10 g/L
Yeast Extract: 5 g/L
Sodium Chloride: 5 g/L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.