Learn More
Boca Scientific Inc LB Broth, Auto-Induction Medium, dehydrated powder, 500 g, beige powder, for protein expression in E. coli, IPTG-inducible promoters, glucose, alpha lactose
Supplier:  Boca Scientific Inc GCM17.0500
Encompass
This LB Broth Auto-Induction Medium is a dehydrated powder formulated for the efficient auto-induction of protein expression in *Escherichia coli*. It simplifies protein expression protocols by eliminating the need for manual monitoring, featuring an optimized blend of glucose and alpha lactose. This product is ideal for high-throughput applications and simultaneous analysis of multiple clones.
Catalog No. NC2736428