Learn More
Growcells LB BROTH LENNOX 1L CS12
Catalog No. 501034798
Encompass_Preferred
Ready to use sterile bacterial culture media, sterile filtered and steam sterilized in TriForest polycarbonate bottles. Recipe/L: Tryptone 10g, Yeast Extract 5g, NaCl 5g pH 7.2
Catalog No. 50-103-4798 Supplier Growcells Supplier No. MBLE706012
Ready to use sterile bacterial culture media, sterile filtered and steam sterilized in TriForest polycarbonate bottles. Recipe/L: Tryptone 10g, Yeast Extract 5g, NaCl 5g pH 7.2