Learn More
Teknova LB Lennox Broth. 1000mL, Sterile.
SDP
Catalog No. 50842375
Encompass
1.0% Tryptone, 0.5% Yeast Extract,0.5% Sodium Chloride
This item is not returnable. View return policy
Catalog No. 50-842-375 Supplier Teknova Supplier No. L5000
1.0% Tryptone, 0.5% Yeast Extract,0.5% Sodium Chloride