Learn More
Sigma Aldrich Fine Chemicals Biosciences LB Broth (Lennox), 250 g
Catalog No. NC1426731
Encompass
Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids and carbohydrates in a low-salt formulation.
Catalog No. NC1426731 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L3022250G
Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids and carbohydrates in a low-salt formulation.