Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc LB Broth (Lennox), Case 10 X 500 mL
SDP

Catalog No. 507517268
Encompass_Preferred

LB Broth, Lennox is a ready-to-use microbial growth medium used for the cultivation of E. coli. Low (5 g/L) salt formulation Lennox is also nutrient rich containing peptides, amino acids, water soluble vitamins and carbohydrates. Useful for molecular genetic studies, as well as maintenance and propagation of bacteriological stocks.

Recomendation: This is the best option for LB Broth if a lower concentration of sodium chloride is desired; as LB Broth, Luria-Bertani Broth has 10.0 g/L NaCl and LB Broth, Lennox contains 5.00 g/L NaCl.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Autoclave Sterilized
Max Shelf Life: 24 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7268 Supplier Quality Biological Inc Supplier No. 340016101CS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LB Broth, Lennox is a ready-to-use microbial growth medium used for the cultivation of E. coli. Low (5 g/L) salt formulation Lennox is also nutrient rich containing peptides, amino acids, water soluble vitamins and carbohydrates. Useful for molecular genetic studies, as well as maintenance and propagation of bacteriological stocks.

Recomendation: This is the best option for LB Broth if a lower concentration of sodium chloride is desired; as LB Broth, Luria-Bertani Broth has 10.0 g/L NaCl and LB Broth, Lennox contains 5.00 g/L NaCl.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Autoclave Sterilized
Max Shelf Life: 24 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.