Learn More
Biotang Inc LB Broth, Lennox Broth, 1KG, P-830, CAS # Sodium Chloride: 7647-14-5, CAS # Yeast Extract: 8013-01-2. For preparation and maintenance of E. coli, 1.0 % Trypton, 0.5% Yeast Extract and 0.5% NaCl, pH, ~7.2.
Catalog No. 50152360 Shop All BIOTANG Inc Products
Encompass_Preferred
LB Broth, Lennox Broth, 1KG, P-830, CAS # Sodium Chloride: 7647-14-5, CAS # Yeast Extract: 8013-01-2. For preparation and maintenance of E. coli, 1.0 % Trypton, 0.5% Yeast Extract and 0.5% NaCl, pH, ~7.2.
Catalog No. 50-152-360 Supplier Biotang Inc Supplier No. BTBB906
LB Broth, Lennox Broth, 1KG, P-830, CAS # Sodium Chloride: 7647-14-5, CAS # Yeast Extract: 8013-01-2. For preparation and maintenance of E. coli, 1.0 % Trypton, 0.5% Yeast Extract and 0.5% NaCl, pH, ~7.2.