Learn More
Sigma Aldrich Fine Chemicals Biosciences LB Broth Lennox Highly ref1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L30226X1KG
Encompass
Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides amino acids and carbohydrates in a low-salt formulation.
Catalog No. 50-291-4624