Learn More
Sigma Aldrich Fine Chemicals Biosciences LB Broth Lennox Highly-ref1KG
Catalog No. 502901817
Encompass
Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides amino acids and carbohydrates in a low-salt formulation.
Catalog No. 50-290-1817 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L30221KG
Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides amino acids and carbohydrates in a low-salt formulation.