Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp LB Broth Lennox Liquid 1 L
SDP

Catalog No. 5000052399
Encompass_Preferred

Sterile Filtered and Autoclaved

LB Broth Lennox liquid is a widely used growth medium designed for cultivating bacteria in life science research settings.

LB Broth Lennox liquid is particularly favored for its compatibility with a diverse range of bacterial strains and its ability to sustain rapid growth rates. With a formulation optimized to minimize salt content, it provides an ideal environment for genetic manipulation and protein expression studies.

Its versatility makes it indispensable for microbiology, molecular biology, and biotechnology applications, serving as a foundational tool for researchers exploring microbial physiology, genetics, and recombinant DNA technologies.

Ingredients:

Tryptone: 10 g/l
Yeast: 5 g/l
Sodium Chloride: 5 g/l

Catalog No. 50-000-52399 Supplier Research Products International Corp Supplier No. L582501000.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sterile Filtered and Autoclaved

LB Broth Lennox liquid is a widely used growth medium designed for cultivating bacteria in life science research settings.

LB Broth Lennox liquid is particularly favored for its compatibility with a diverse range of bacterial strains and its ability to sustain rapid growth rates. With a formulation optimized to minimize salt content, it provides an ideal environment for genetic manipulation and protein expression studies.

Its versatility makes it indispensable for microbiology, molecular biology, and biotechnology applications, serving as a foundational tool for researchers exploring microbial physiology, genetics, and recombinant DNA technologies.

Ingredients:

Tryptone: 10 g/l
Yeast: 5 g/l
Sodium Chloride: 5 g/l

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.