Learn More
Alkali Scientific LB Broth [Lennox] Powder, 1 Kg
SDP
Supplier:  Alkali Scientific THL001CP
Encompass_Preferred
LB Broth (Lennox) Powder is a widely used microbiological medium for the growth of Escherichia coli and other microorganisms. This 1kg powder formulation is ideal for laboratories that require a simple, reliable medium for culturing bacteria. The Lennox formulation of LB Broth is preferred for applications such as cloning, plasmid propagation, and general bacterial culture. It provides the necessary nutrients, including amino acids and salts, to support rapid bacterial growth. This medium is commonly used in molecular biology and biochemistry labs for routine bacterial cultivation and is suitable for large-scale culture when needed.
Catalog No. 50-006-61255