Learn More
Alkali Scientific LB Broth [Lennox] Powder, 10 Kg
SDP
Catalog No. 5000660511
Encompass_Preferred
LB Broth (Lennox) Powder, 10kg, is designed for high-volume applications in microbiology and molecular biology labs. This high-performance medium supports the growth of a wide range of bacterial species, particularly Escherichia coli, which is commonly used in genetic engineering and cloning studies. The Lennox formulation offers a balanced nutrient profile that ensures optimal bacterial growth while maintaining the simplicity and cost-effectiveness of standard LB media. This medium is widely used in recombinant DNA research, protein expression, and general bacterial culture for laboratory applications. The powder form provides flexibility and convenience in large-scale preparations.
Catalog No. 50-006-60511 Supplier Alkali Scientific Supplier No. THL002CP
LB Broth (Lennox) Powder, 10kg, is designed for high-volume applications in microbiology and molecular biology labs. This high-performance medium supports the growth of a wide range of bacterial species, particularly Escherichia coli, which is commonly used in genetic engineering and cloning studies. The Lennox formulation offers a balanced nutrient profile that ensures optimal bacterial growth while maintaining the simplicity and cost-effectiveness of standard LB media. This medium is widely used in recombinant DNA research, protein expression, and general bacterial culture for laboratory applications. The powder form provides flexibility and convenience in large-scale preparations.