Learn More
Alkali Scientific LB Broth [Lennox] Powder, 5 Kg
SDP
Catalog No. 5000661038
Encompass_Preferred
LB Broth (Lennox) Powder (5kg) is designed for high-throughput laboratories that require a large volume of growth medium for bacterial culture. The Lennox formulation is particularly well-suited for culturing fast-growing bacteria like Escherichia coli, commonly used in genetic research, protein production, and plasmid propagation. The high-quality nutrients and balanced salt content in this medium ensure robust bacterial growth and reliable results. This bulk 5kg powder format is perfect for laboratories with high-volume culture needs, providing cost efficiency while maintaining the medium's integrity and quality for large-scale microbial applications.
Catalog No. 50-006-61038 Supplier Alkali Scientific Supplier No. THL005CP
LB Broth (Lennox) Powder (5kg) is designed for high-throughput laboratories that require a large volume of growth medium for bacterial culture. The Lennox formulation is particularly well-suited for culturing fast-growing bacteria like Escherichia coli, commonly used in genetic research, protein production, and plasmid propagation. The high-quality nutrients and balanced salt content in this medium ensure robust bacterial growth and reliable results. This bulk 5kg powder format is perfect for laboratories with high-volume culture needs, providing cost efficiency while maintaining the medium's integrity and quality for large-scale microbial applications.